Anti-MUC20 Rabbit Polyclonal Antibody
Catalog # 77743-642
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MUC20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8N307
- Antigen Synonyms:MUC 20|MUC-20|MUC20_HUMAN|Mucin 20|cell surface associated|Mucin-20|Transmembrane mucin MUC20S
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transmembrane mucin MUC20S
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MUC20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human