Anti-IL16 Rabbit Polyclonal Antibody
77740-787
New Product
- Antigen Symbol:IL16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q14005
- Antigen Synonyms:HGNC:5980|HsT19289|IL-16|IL16_HUMAN|Interleukin 16 precursor|Interleukin-16|LCF|Lymphocyte chemoattractant factor|Neuronal interleukin 16|NprIL 16|PrProinterleukin 16
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:HGNC:5980
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: IL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human