Anti-SIRT3 Rabbit Polyclonal Antibody
Catalog # 77736-798
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SIRT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NTG7
- Antigen Synonyms:hSIRT 3|hMitochondrial nicotinamide adenine dinucleotide dependent deacetylase|NAD dependent deacetylase sirtuin 3 mitochondrial|NAD-dependent protein deacetylase sirtuin-3, mitochondrial|Regulatory protein SIR2 homolog 3|Silent mating type information regulation 2 S.cerevisiae homolog 3|Sir 2 like 3|SIR 2 like protein 3|SIR 3|SIR2 L3|Sir2 like 3|SIR2 like protein 3|SIR2-like protein 3|SIR2L3|SIR3_HUMAN|SIRT 3|Sirtuin 3|Sirtuin silent mating type information regulation 2 homolog 3 (S. cerevisiae)|Sirtuin type 3|Sirtuin3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:hSIRT 3
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:5000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human