Anti-DYNLL1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DYNLL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-884
- 77742-053
- Antigen Symbol:
- DYNLL1
- DYNLL1
- Antigen Synonyms:
- 8 kDa dynein light chain|8kDLC|Cytoplasmic dynein light polypeptide|DLC1|DLC8|DNCL1|DNCLC1|DYL1_HUMAN|Dynein|cytoplasmic|light chain 1|Dynein light chain 1 cytoplasmic|Dynein light chain 1|Dynein light chain LC8 type 1|Dynein light chain LC8-type 1|light polypeptide 1|light chain|LC8-type 1|HDLC1|LC8|LC8a|MGC126137|MGC126138|MGC72986|PIN|Protein inhibitor of neuronal nitric oxide synthase|Protein inhibitor of neuronal NOS
- 8 kDa dynein light chain|8kDLC|Cytoplasmic dynein light polypeptide|DLC1|DLC8|DNCL1|DNCLC1|DYL1_HUMAN|Dynein|cytoplasmic|light chain 1|Dynein light chain 1 cytoplasmic|Dynein light chain 1|Dynein light chain LC8 type 1|Dynein light chain LC8-type 1|light polypeptide 1|light chain|LC8-type 1|HDLC1|LC8|LC8a|MGC126137|MGC126138|MGC72986|PIN|Protein inhibitor of neuronal nitric oxide synthase|Protein inhibitor of neuronal NOS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P63167
- P63167
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dynein light chain 1, cytoplasmic protein (1-89AA)
- Recombinant Human Dynein light chain 1, cytoplasmic protein (1-89AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together