Anti-FPR2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FPR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-595
- 77737-489
- Antigen Symbol:
- FPR2
- FPR2
- Antigen Synonyms:
- ALXR| FMLP-R-I| FMLP-R-II| FMLP-related receptor I| FMLPX| Formyl peptide receptor 2| Formyl peptide receptor related| Formyl peptide receptor-like 1| FPR2| FPR2_HUMAN| FPR2A| FPRH1| FPRH2| FPRL1| HM63| Lipoxin A4 receptor| LXA4 receptor| LXA4R| N-formyl peptide receptor 2| RFP
- ALXR|FMLP-R-I|FMLP-R-II|FMLP-related receptor I|FMLPX|Formyl peptide receptor 2|Formyl peptide receptor related|Formyl peptide receptor-like 1|FPR2|FPR2_HUMAN|FPR2A|FPRH1|FPRH2|FPRL1|HM63|Lipoxin A4 receptor|LXA4 receptor|LXA4R|N-formyl peptide receptor 2|RFP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25090
- P25090
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human N-formyl peptide receptor 2 protein (307-351AA)
- Recombinant Human N-formyl peptide receptor 2 protein (307-351AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together