Anti-GOLM1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GOLM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-259
- 77738-667
- Antigen Symbol:
- GOLM1
- GOLM1
- Antigen Synonyms:
- bA379P1.3|C9orf155|Chromosome 9 open reading frame 155|Golgi membrane protein 1|Golgi membrane protein GP73|Golgi phosphoprotein 2|Golgi protein 73 kD|Golgi protein 73kD|GOLM 1|GOLM1_HUMAN|GOLPH 2|GOLPH2|GP 73|GP73|PSEC0257
- bA379P1.3|C9orf155|Chromosome 9 open reading frame 155|Golgi membrane protein 1|Golgi membrane protein GP73|Golgi phosphoprotein 2|Golgi protein 73 kD|Golgi protein 73kD|GOLM 1|GOLM1_HUMAN|GOLPH 2|GOLPH2|GP 73|GP73|PSEC0257
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NBJ4
- Q8NBJ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Golgi membrane protein 1 protein (112-401AA)
- Recombinant Human Golgi membrane protein 1 protein (112-401AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together