Anti-MDH1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MDH1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77742-926
- 77859-604
- Antigen Symbol:
- MDH1
- MDH1
- Antigen Synonyms:
- cytoplasmic|Cytosolic malate dehydrogenase|Diiodophenylpyruvate reductase|Malate dehydrogenase 1, NAD (soluble)|Malate dehydrogenase|Malate dehydrogenase cytoplasmic|MDH s|mdh1|MDHA|MDHC_HUMAN|MDHs|MGC:1375|MOR2|Soluble malate dehydrogenase
- cytoplasmic|Cytosolic malate dehydrogenase|Diiodophenylpyruvate reductase|Malate dehydrogenase 1, NAD (soluble)|Malate dehydrogenase|Malate dehydrogenase cytoplasmic|MDH s|mdh1|MDHA|MDHC_HUMAN|MDHs|MGC:1375|MOR2|Soluble malate dehydrogenase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P40925
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Malate dehydrogenase, cytoplasmic protein (1-334AA)
- Recombinant Human Malate dehydrogenase, cytoplasmic protein (1-334AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together