Anti-IFIT3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IFIT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-640
- 77857-849
- Antigen Symbol:
- IFIT3
- IFIT3
- Antigen Synonyms:
- CIG 49|CIG49|GARG 49|IFI-60K|IFI60|IFI60K|IFIT-3|IFIT-4|Ifit3|IFIT3_HUMAN|IFIT4|Interferon induced 60 kDa protein|Interferon induced protein 60|Interferon induced protein with tetratricopeptide repeats 3|Interferon-induced 60 kDa protein|Interferon-induced protein with tetratricopeptide repeats 3|Interferon-induced protein with tetratricopeptide repeats 4|IRG2|ISG-60|ISG60|P60|Retinoic acid induced gene G protein|Retinoic acid-induced gene G protein|RIG G|RIG-G|RIGG|RP11-149I23.4
- CIG 49|CIG49|GARG 49|IFI-60K|IFI60|IFI60K|IFIT-3|IFIT-4|Ifit3|IFIT3_HUMAN|IFIT4|Interferon induced 60 kDa protein|Interferon induced protein 60|Interferon induced protein with tetratricopeptide repeats 3|Interferon-induced 60 kDa protein|Interferon-induced protein with tetratricopeptide repeats 3|Interferon-induced protein with tetratricopeptide repeats 4|IRG2|ISG-60|ISG60|P60|Retinoic acid induced gene G protein|Retinoic acid-induced gene G protein|RIG G|RIG-G|RIGG|RP11-149I23.4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14879
- O14879
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Interferon-induced protein with tetratricopeptide repeats 3 protein (1-490AA)
- Recombinant Human Interferon-induced protein with tetratricopeptide repeats 3 protein (1-490AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together