Anti-KRT4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-887
- 77858-749
- Antigen Symbol:
- KRT4
- KRT4
- Antigen Synonyms:
- CK 4|CK-4|CK4|CYK4|Cytokeratin 4|Cytokeratin-4|Cytokeratin4|FLJ31692|K2C4_HUMAN|K4|Keratin 4|Keratin|Keratin type II cytoskeletal 4|Keratin-4|Keratin4|KRT 4|Krt4|type II cytoskeletal 4|Type-II keratin Kb4
- CK 4|CK-4|CK4|CYK4|Cytokeratin 4|Cytokeratin-4|Cytokeratin4|FLJ31692|K2C4_HUMAN|K4|Keratin 4|Keratin|Keratin type II cytoskeletal 4|Keratin-4|Keratin4|KRT 4|Krt4|type II cytoskeletal 4|Type-II keratin Kb4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P19013
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Keratin, type II cytoskeletal 4 protein (1-534AA)
- Recombinant Human Keratin, type II cytoskeletal 4 protein (1-534AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together