Anti-EPHB3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-011
- 77854-813
- Antigen Symbol:
- EPHB3
- EPHB3
- Antigen Synonyms:
- Cek10|EK2|Embryonic kinase 2|EPH Like Tyrosine Kinase 2|EPH receptor B3|EPH-like kinase 2|EPHB3_HUMAN|Ephrin receptor Ephrin type B receptor 3|Ephrin type-B receptor 3|ETK2|hEK2|Human Embryo Kinase 2|Mdk5|Sek4|TYRO6|Tyrosine protein kinase receptor HEK2|Tyrosine protein kinase TYRO6|Tyrosine-protein kinase TYRO6
- Cek10|EK2|Embryonic kinase 2|EPH Like Tyrosine Kinase 2|EPH receptor B3|EPH-like kinase 2|EPHB3_HUMAN|Ephrin receptor Ephrin type B receptor 3|Ephrin type-B receptor 3|ETK2|hEK2|Human Embryo Kinase 2|Mdk5|Sek4|TYRO6|Tyrosine protein kinase receptor HEK2|Tyrosine protein kinase TYRO6|Tyrosine-protein kinase TYRO6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54753
- P54753
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ephrin type-B receptor 3 protein (261-390AA)
- Recombinant Human Ephrin type-B receptor 3 protein (261-390AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together