Anti-HNRNPA2B1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: HNRNPA2B1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-225
- 77857-518
- Antigen Symbol:
- HNRNPA2B1
- HNRNPA2B1
- Antigen Synonyms:
- Heterogeneous nuclear ribonucleoprotein A2|Heterogeneous nuclear ribonucleoprotein A2/B1|Heterogeneous nuclear ribonucleoprotein B1|Heterogeneous nuclear ribonucleoproteins A2/B1|hnRNP A2 / hnRNP B1|hnRNP A2|hnRNP A2/B1|hnRNP B1|hnRNP-A2|hnRNP-B1|hnRNPA2|hnRNPB1|HNRPA2|HNRPA2B1|HNRPB1|Nuclear ribonucleoprotein particle A2 protein|RNP A2|RNP B1|RNP-A2|RNP-B1|RNPA2|RNPB1|ROA2_HUMAN|SNRPB1
- Heterogeneous nuclear ribonucleoprotein A2|Heterogeneous nuclear ribonucleoprotein A2/B1|Heterogeneous nuclear ribonucleoprotein B1|Heterogeneous nuclear ribonucleoproteins A2/B1|hnRNP A2 / hnRNP B1|hnRNP A2|hnRNP A2/B1|hnRNP B1|hnRNP-A2|hnRNP-B1|hnRNPA2|hnRNPB1|HNRPA2|HNRPA2B1|HNRPB1|Nuclear ribonucleoprotein particle A2 protein|RNP A2|RNP B1|RNP-A2|RNP-B1|RNPA2|RNPB1|ROA2_HUMAN|SNRPB1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P22626
- P22626
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Heterogeneous nuclear ribonucleoproteins A2/B1 protein (1-249AA)
- Recombinant Human Heterogeneous nuclear ribonucleoproteins A2/B1 protein (1-249AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together