Anti-HIP1R Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HIP1R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-859
- 77739-697
- Antigen Symbol:
- HIP1R
- HIP1R
- Antigen Synonyms:
- FLJ14000|FLJ27022|HIP-12|Hip1 related|HIP1-related protein|HIP12|HIP1R_HUMAN|HIP3|Huntingtin Interacting Protein 1 Related|Huntingtin interacting protein 12|Huntingtin-interacting protein 1-related protein|Huntingtin-interacting protein 12|ILWEQ|KIAA0655|MGC47513
- FLJ14000|FLJ27022|HIP-12|Hip1 related|HIP1-related protein|HIP12|HIP1R_HUMAN|HIP3|Huntingtin Interacting Protein 1 Related|Huntingtin interacting protein 12|Huntingtin-interacting protein 1-related protein|Huntingtin-interacting protein 12|ILWEQ|KIAA0655|MGC47513
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75146
- O75146
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Huntingtin-interacting protein 1-related protein (304-487AA)
- Recombinant Human Huntingtin-interacting protein 1-related protein (304-487AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together