Anti-GOSR2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GOSR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77738-684
- 77856-267
- Antigen Symbol:
- GOSR2
- Antigen Synonyms:
- 2310032N09Rik|27 kDa Golgi SNARE protein|Bos1|EPM6|Golgi SNAP receptor complex member 2|Golgi SNARE|Gosr2|GOSR2_HUMAN|Gs27|Membrin|SNARE
- 2310032N09Rik|27 kDa Golgi SNARE protein|Bos1|EPM6|Golgi SNAP receptor complex member 2|Golgi SNARE|Gosr2|GOSR2_HUMAN|Gs27|Membrin|SNARE
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- O14653
- O14653
- Isotype:
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Golgi SNAP receptor complex member 2 protein (1-190AA)
- Recombinant Human Golgi SNAP receptor complex member 2 protein (1-190AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together