Order Entry
United States
ContactUsLinkComponent
Anti-KIR2DL2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-KIR2DL2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-KIR2DL2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77741-694
  • 77858-604
  • Antigen Symbol:
  • KIR2DL2
  • KIR2DL2
  • Antigen Synonyms:
  • KIR2DL2|CD158B1|NKAT6Killer cell immunoglobulin-like receptor 2DL2|CD158 antigen-like family member B1|MHC class I NK cell receptor|Natural killer-associated transcript 6|NKAT-6|p58 natural killer cell receptor clone CL-43|p58 NK receptor CL-43|CD antigen CD158b1
  • KIR2DL2|CD158B1|NKAT6Killer cell immunoglobulin-like receptor 2DL2|CD158 antigen-like family member B1|MHC class I NK cell receptor|Natural killer-associated transcript 6|NKAT-6|p58 natural killer cell receptor clone CL-43|p58 NK receptor CL-43|CD antigen CD158b1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P43627
  • P43627
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Killer cell immunoglobulin-like receptor 2DL2 protein (22-245AA)
  • Recombinant Human Killer cell immunoglobulin-like receptor 2DL2 protein (22-245AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together