Anti-LDHB Rabbit Polyclonal Antibody
Catalog # 77742-044
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:LDHB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P07195
- Antigen Synonyms:Epididymis secretory protein Li 281|HEL S 281|L lactate dehydrogenase B chain|L-lactate dehydrogenase B chain|Lactate Dehydrogenase B|Lactate dehydrogenase H chain|LDH B|LDH H|LDH heart subunit|LDH-B|LDH-H|LDHB|LDHB_HUMAN|LDHBD|LDHH|Renal carcinoma antigen NY REN 46|Renal carcinoma antigen NY-REN-46|TRG-5|TRG5
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TRG5
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: LDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat