Anti-MYBL2 Rabbit Polyclonal Antibody
Catalog # 77739-644
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MYBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P10244
- Antigen Synonyms:B-Myb|BMyB|MGC15600|MYB L2|Myb related protein B|Myb-like protein 2|Myb-related protein B|MybB|MYBB_HUMAN|MYBL 2|v myb avian myeloblastosis viral oncogene homolog like 2|v myb myeloblastosis viral oncogene homolog (avian) like 2|v myb myeloblastosis viral oncogene homolog like 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:B-Myb
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:40000
Type:
Antigen: MYBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat