Anti-KDM5A Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: KDM5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-478
- 77741-544
- Antigen Symbol:
- KDM5A
- KDM5A
- Antigen Synonyms:
- Histone demethylase JARID1A|JARID1A|Jumonji/ARID domain containing protein 1A|Jumonji/ARID domain-containing protein 1A|KDM5A_HUMAN|Lysine-specific demethylase 5A|RBBP-2|RBBP2|RBP2|Retinoblastoma binding protein 2|Retinoblastoma-binding protein 2
- Histone demethylase JARID1A|JARID1A|Jumonji/ARID domain containing protein 1A|Jumonji/ARID domain-containing protein 1A|KDM5A_HUMAN|Lysine-specific demethylase 5A|RBBP-2|RBBP2|RBP2|Retinoblastoma binding protein 2|Retinoblastoma-binding protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P29375
- P29375
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysine-specific demethylase 5A protein (437-603AA)
- Recombinant Human Lysine-specific demethylase 5A protein (437-603AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together