Anti-USF2 Rabbit Polyclonal Antibody
Catalog # 77740-712
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:USF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15853
- Antigen Synonyms:bHLHb12|Class B basic helix-loop-helix protein 12|FIP|FOS interacting protein|FOS-interacting protein|Major late transcription factor 2|Upstream stimulatory factor 2|Upstream transcription factor 2|USF 2|USF2_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:bHLHb12
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA:1:20000
Type:
Antigen: USF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat