Anti-LPAR4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LPAR4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-095
- 77742-307
- Antigen Symbol:
- LPAR4
- LPAR4
- Antigen Synonyms:
- 5730485F04Rik|G protein coupled receptor 23|G-protein coupled receptor 23|GPR23|LPA receptor 4|LPA-4|LPA4|LPAR4_HUMAN|Lysophosphatidic acid receptor 4|Lysophosphatidic Acid Receptor P2Y9|P2RY9|P2Y purinoceptor 9|P2Y5 like|P2Y5-like receptor|P2Y9|Purinergic receptor 9|Purinergic receptor|P2Y|G-protein coupled|9
- P2Y5-like receptor|GPR23|G protein coupled receptor 23|P2Y9|Lysophosphatidic acid receptor 4|P2RY9|LPA-4|G-protein coupled|G-protein coupled receptor 23|LPA4|Purinergic receptor|P2Y|P2Y5 like|9|LPAR4_HUMAN|Lysophosphatidic Acid Receptor P2Y9|P2Y purinoceptor 9|LPA receptor 4|Purinergic receptor 9|5730485F04Rik
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99677
- Q99677
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysophosphatidic acid receptor 4 protein (1-43AA)
- Recombinant Human Lysophosphatidic acid receptor 4 protein (1-43AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together