Anti-NEU4 Rabbit Polyclonal Antibody
Catalog # 77744-327
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NEU4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8WWR8
- Antigen Synonyms:LP5125Sialidase-4|EC 3.2.1.18|N-acetyl-alpha-neuraminidase 4
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:N-acetyl-alpha-neuraminidase 4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NEU4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human