Order Entry
United States
ContactUsLinkComponent
 
Anti-LPCAT2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-LPCAT2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77859-103
  • 77742-316
  • Antigen Symbol:
  • LPCAT2
  • LPCAT2
  • Antigen Synonyms:
  • 1 acylglycerophosphocholine O acyltransferase|1 alkylglycerophosphocholine O acetyltransferase|1-acylglycerophosphocholine O-acyltransferase|1-alkylglycerophosphocholine O-acetyltransferase|A330042H22|Acetyl CoA:lyso PAF acetyltransferase|Acetyl-CoA:lyso-PAF acetyltransferase|Acetyl-CoA:lyso-platelet-activating factor acetyltransferase|acyl CoA:lysophosphatidylcholine acyltransferase 2|Acyl-CoA:lysophosphatidylcholine acyltransferase member 1|Acyltransferase like 1|Acyltransferase like 1A|Acyltransferase-like 1|AYTL1|Aytl1a|DKFZp686H22112|FLJ20481|lpafat1|LPC acyltransferase 2|LPCAT-2|Lyso PAF acetyltransferase|Lyso platelet activating factor (PAF) acetyltransferase|Lyso-PAF acetyltransferase|LysoPAFAT|LysoPAFAT/LysoPC acyltransferase 2|Lysophosphatidylcholine acyltransferase 2|OTTMUSP00000029641|PCAT2_HUMAN|RGD1563994
  • 1 acylglycerophosphocholine O acyltransferase|1 alkylglycerophosphocholine O acetyltransferase|1-acylglycerophosphocholine O-acyltransferase|1-alkylglycerophosphocholine O-acetyltransferase|A330042H22|Acetyl CoA:lyso PAF acetyltransferase|Acetyl-CoA:lyso-PAF acetyltransferase|Acetyl-CoA:lyso-platelet-activating factor acetyltransferase|acyl CoA:lysophosphatidylcholine acyltransferase 2|Acyl-CoA:lysophosphatidylcholine acyltransferase member 1|Acyltransferase like 1|Acyltransferase like 1A|Acyltransferase-like 1|AYTL1|Aytl1a|DKFZp686H22112|FLJ20481|lpafat1|LPC acyltransferase 2|LPCAT-2|Lyso PAF acetyltransferase|Lyso platelet activating factor (PAF) acetyltransferase|Lyso-PAF acetyltransferase|LysoPAFAT|LysoPAFAT/LysoPC acyltransferase 2|Lysophosphatidylcholine acyltransferase 2|OTTMUSP00000029641|PCAT2_HUMAN|RGD1563994
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q7L5N7
  • Q7L5N7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Lysophosphatidylcholine acyltransferase 2 protein (442-544AA)
  • Recombinant Human Lysophosphatidylcholine acyltransferase 2 protein (442-544AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together