Anti-ZDHHC20 Rabbit Polyclonal Antibody
Catalog # 77736-965
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZDHHC20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q5W0Z9
- Antigen Synonyms:4933421L13Rik|DHHC-20|Probable palmitoyltransferase ZDH20_HUMAN|Zinc finger DHHC domain-containing protein 20|Zinc finger DHHC type containing 20
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:4933421L13Rik
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000
Type:
Antigen: ZDHHC20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat