Anti-acpP Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: acpP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Specifications
- Catalog No:
- 77744-161
- 77744-162
- Antigen Symbol:
- acpP
- acpP
- Antigen Synonyms:
- hmrB|SAV1232Acyl carrier protein|ACP
- hmrB|SAV1232Acyl carrier protein|ACP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Staphylococcus aureus
- Staphylococcus aureus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0A001
- P0A001
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Staphylococcus aureus Acyl carrier protein (1-77AA)
- Recombinant Staphylococcus aureus Acyl carrier protein (1-77AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together