Anti-MLYCD Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MLYCD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-836
- 77743-234
- Antigen Symbol:
- MLYCD
- MLYCD
- Antigen Synonyms:
- DCMC_HUMAN|hMCD|Malonyl CoA decarboxylase|Malonyl CoA decarboxylase mitochondrial|Malonyl coenzyme A decarboxylase|Malonyl-CoA decarboxylase|MCD|MGC59795|mitochondrial|Mlycd
- DCMC_HUMAN|hMCD|Malonyl CoA decarboxylase|Malonyl CoA decarboxylase mitochondrial|Malonyl coenzyme A decarboxylase|Malonyl-CoA decarboxylase|MCD|MGC59795|mitochondrial|Mlycd
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95822
- O95822
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Malonyl-CoA decarboxylase, mitochondrial protein (40-493AA)
- Recombinant human Malonyl-CoA decarboxylase, mitochondrial protein (40-493AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together