Anti-FCGR2C Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: FCGR2C
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-982
- 77855-252
- Antigen Symbol:
- FCGR2C
- FCGR2C
- Antigen Synonyms:
- CD32| CDw32| Fc-gamma RII-c| Fc-gamma-RIIc| FCG2C_HUMAN| FcRII-c| IgG Fc receptor II-c| Low affinity immunoglobulin gamma Fc region receptor II-c
- CD32|CDw32|Fc-gamma RII-c|Fc-gamma-RIIc|FCG2C_HUMAN|FcRII-c|IgG Fc receptor II-c|Low affinity immunoglobulin gamma Fc region receptor II-c
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31995
- P31995
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Low affinity immunoglobulin gamma Fc region receptor II-c protein (43-223AA)
- Recombinant Human Low affinity immunoglobulin gamma Fc region receptor II-c protein (43-223AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together