Anti-Map2k4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Map2k4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77742-646
- 77859-375
- Antigen Symbol:
- Map2k4
- Map2k4
- Antigen Synonyms:
- Map2k4|Jnkk1|Mek4|Mkk4|Prkmk4|Sek1|Serk1|Skk1|Dual specificity mitogen-activated protein kinase kinase 4|MAP kinase kinase 4|MAPKK 4|EC 2.7.12.2|C-JUN N-terminal kinase kinase 1|JNK kinase 1|JNKK 1|JNK-activating kinase 1|MAPK/ERK kinase 4|MEK 4|SAPK/ERK kinase 1|SEK1
- Map2k4| Jnkk1| Mek4| Mkk4| Prkmk4| Sek1| Serk1| Skk1| Dual specificity mitogen-activated protein kinase kinase 4| MAP kinase kinase 4| MAPKK 4| EC 2.7.12.2| C-JUN N-terminal kinase kinase 1| JNK kinase 1| JNKK 1| JNK-activating kinase 1| MAPK/ERK kinase 4| MEK 4| SAPK/ERK kinase 1| SEK1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P47809
- P47809
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse Dual specificity mitogen-activated protein kinase kinase 4 protein (2-397AA)
- Recombinant Mouse Dual specificity mitogen-activated protein kinase kinase 4 protein (2-397AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together