Anti-RAB37 Rabbit Polyclonal Antibody
Catalog # 77737-992
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RAB37
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96AX2
- Antigen Synonyms:B230331O03Rik| B230354I04Rik| FLJ30284| FLJ32507| GTPase MGC157622| OTTMUSP00000003775| RAB 37| Rab37 like| RAB37 member RAS oncogene family| RAB37_HUMAN| Ras related protein Rab 37| Ras related protein Ras-related protein Rab-37| RP23-342L24.2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:B230331O03Rik
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: RAB37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human