Anti-FLI1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FLI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-465
- 77737-275
- Antigen Symbol:
- FLI1
- FLI1
- Antigen Synonyms:
- ERGB transcription factor|Ewing Sarcoma breakpoint region 2|EWSR 2|EWSR2|FLI 1|FLI 1 proto oncogene|Fli-1 proto-oncogene|ETS transcription factor|FLI1 EWS fusion gene|FLI1 proto oncogene|FLI1_HUMAN|Friend leukemia integration 1 (FLI1) transcription factor|Friend leukemia integration 1 transcription factor|Friend leukemia virus integration 1|Proto-oncogene Fli-1|Sarcoma breakpoint region 2 (EWSR2)|SIC 1|SIC1|Transcription factor ERGB|Viral integration region mouse|homolog of
- SIC1|FLI1 proto oncogene|FLI1 EWS fusion gene|Friend leukemia virus integration 1|FLI 1|EWSR 2|Viral integration region SIC 1|ERGB transcription factor|homolog of|ETS transcription factor|mouse|Sarcoma breakpoint region 2 (EWSR2)|EWSR2|Ewing Sarcoma breakpoint region 2|Friend leukemia integration 1 transcription factor|Proto-oncogene Fli-1|Transcription factor ERGB|FLI 1 proto oncogene|FLI1_HUMAN|Friend leukemia integration 1 (FLI1) transcription factor|Fli-1 proto-oncogene
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01543
- Q01543
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Friend leukemia integration 1 transcription factor protein (205-331AA)
- Recombinant Human Friend leukemia integration 1 transcription factor protein (205-331AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together