Anti-HOXB4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXB4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-584
- 77740-302
- Antigen Symbol:
- HOXB4
- HOXB4
- Antigen Synonyms:
- Homeo box 2F|Homeo box B4|Homeobox 2F|Homeobox B4|Homeobox protein Hox B4|Homeobox protein Hox-2.6|Homeobox protein Hox-2F|Homeobox protein Hox-B4|Homeobox protein HoxB4|HOX 2|Hox 2.6|Hox 2F|HOX B4|HOX2|Hox2.6|Hox2F|HOXB 4|hoxb4|HXB4_HUMAN
- Homeo box 2F|Homeo box B4|Homeobox 2F|Homeobox B4|Homeobox protein Hox B4|Homeobox protein Hox-2.6|Homeobox protein Hox-2F|Homeobox protein Hox-B4|Homeobox protein HoxB4|HOX 2|Hox 2.6|Hox 2F|HOX B4|HOX2|Hox2.6|Hox2F|HOXB 4|hoxb4|HXB4_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17483
- P17483
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-B4 protein (88-166AA)
- Recombinant Human Homeobox protein Hox-B4 protein (88-166AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together