Anti-LAIR2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LAIR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-809
- 77741-950
- Antigen Symbol:
- LAIR2
- LAIR2
- Antigen Synonyms:
- CD306|CD306 antigen|LAIR 2|LAIR-2|LAIR2_HUMAN|Leukocyte associated Ig like receptor 2|Leukocyte associated immunoglobulin like receptor 2|Leukocyte-associated immunoglobulin-like receptor 2|MGC71634|OTTHUMP00000068389|OTTHUMP00000068390
- CD306|CD306 antigen|LAIR 2|LAIR-2|LAIR2_HUMAN|Leukocyte associated Ig like receptor 2|Leukocyte associated immunoglobulin like receptor 2|Leukocyte-associated immunoglobulin-like receptor 2|MGC71634|OTTHUMP00000068389|OTTHUMP00000068390
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ISS4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 2 protein (22-152AA)
- Recombinant Human Leukocyte-associated immunoglobulin-like receptor 2 protein (22-152AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together