Anti-AGAP2 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:50 to 1:200
Type:
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77859-807
- 77743-188
- Antigen Symbol:
- AGAP2
- Antigen Synonyms:
- AGAP-2|Agap2|Agap2 ArfGAP with GTPase domain|ankyrin repeat and PH domain 2|AGAP2_HUMAN|ANK repeat and PH domains 2|Arf GAP with GTP binding protein like|Arf GAP with GTP-binding protein-like|ankyrin repeat and pleckstrin homology domains 2|Arf-GAP with GTPase|ANK repeat and PH domain-containing protein 2|ArfGAP with GTPase domain|Centaurin gamma 1|centaurin gamma1|Centaurin-gamma-1|Centg1|cnt g1|Cnt-g1|Cnt2|FLJ16430|GGAP2|GTP binding and GTPase activating protein 2|GTP-binding and GTPase activating protein 2|GTP-binding and GTPase-activating protein 2|KIAA0167|mKIAA0167|Nuclear GTPase PIKE|Phosphatidylinositol 3-kinase enhancer|Phosphoinositide 3 kinase enhancer|PI3K ENHANCER|PIKE
- AGAP-2|Agap2|Agap2 ArfGAP with GTPase domain|ankyrin repeat and PH domain 2|AGAP2_HUMAN|ANK repeat and PH domains 2|Arf GAP with GTP binding protein like|Arf GAP with GTP-binding protein-like|ankyrin repeat and pleckstrin homology domains 2|Arf-GAP with GTPase|ANK repeat and PH domain-containing protein 2|ArfGAP with GTPase domain|Centaurin gamma 1|centaurin gamma1|Centaurin-gamma-1|Centg1|cnt g1|Cnt-g1|Cnt2|FLJ16430|GGAP2|GTP binding and GTPase activating protein 2|GTP-binding and GTPase activating protein 2|GTP-binding and GTPase-activating protein 2|KIAA0167|mKIAA0167|Nuclear GTPase PIKE|Phosphatidylinositol 3-kinase enhancer|Phosphoinositide 3 kinase enhancer|PI3K ENHANCER|PIKE
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q99490
- Q99490
- Isotype:
- IgG
- Western Blot:
- Yes
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Synthetic peptide of Human AGAP2
- Synthetic peptide of Human AGAP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together