Anti-NOMO2 Rabbit Polyclonal Antibody
Catalog # 77744-639
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NOMO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q5JPE7
- Antigen Synonyms:NOMO2|Nodal modulator 2|pM5 protein 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:pM5 protein 2
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NOMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human