Anti-MYF5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYF5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-183
- 77743-715
- Antigen Symbol:
- MYF5
- MYF5
- Antigen Synonyms:
- bHLHc2|Class C basic helix loop helix protein 2|Class C basic helix-loop-helix protein 2|Myf 5|Myf-5|MYF5_HUMAN|Myogenic factor 5
- bHLHc2|Class C basic helix loop helix protein 2|Class C basic helix-loop-helix protein 2|Myf 5|Myf-5|MYF5_HUMAN|Myogenic factor 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P13349
- P13349
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Myogenic factor 5 protein (1-255AA)
- Recombinant Human Myogenic factor 5 protein (1-255AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together