Anti-FEN1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FEN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: A. thaliana
Specifications
- Catalog No:
- 77737-046
- 77855-291
- Antigen Symbol:
- FEN1
- FEN1
- Antigen Synonyms:
- At5g26680|F21E10.3Flap endonuclease 1|FEN-1|EC 3.1.-.-|Flap structure-specific endonuclease 1
- At5g26680|F21E10.3Flap endonuclease 1|FEN-1|EC 3.1.-.-|Flap structure-specific endonuclease 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- A. thaliana
- A. thaliana
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O65251
- O65251
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Arabidopsis thaliana flap endonuclease 1 protein (85-383AA)
- Recombinant Arabidopsis thaliana Flap endonuclease 1 protein (85-383AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together