Anti-NR2C2AP Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NR2C2AP
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77744-938
- 77744-937
- Antigen Symbol:
- NR2C2AP
- NR2C2AP
- Antigen Synonyms:
- NR2CA_HUMAN| Nuclear receptor 2C2 associated protein| Nuclear receptor 2C2-associated protein| Repressor for TR4 transactivation| TR4 orphan receptor associated 16 kDa protein| TR4 orphan receptor associated protein TRA16| TR4 orphan receptor-associated 16 kDa protein
- NR2CA_HUMAN| Nuclear receptor 2C2 associated protein| Nuclear receptor 2C2-associated protein| Repressor for TR4 transactivation| TR4 orphan receptor associated 16 kDa protein| TR4 orphan receptor associated protein TRA16| TR4 orphan receptor-associated 16 kDa protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WQ0
- Q86WQ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Nuclear receptor 2C2-associated protein (7-139AA)
- Nuclear receptor 2C2-associated protein (7-139AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together