Anti-IER3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50 to 1:200
Type:
Antigen: IER3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-619
- 77857-830
- Antigen Symbol:
- IER3
- IER3
- Antigen Synonyms:
- DIF2|Differentiation-dependent gene 2 protein|GLY96|IER3|IEX1_HUMAN|IEX1L|Immediate early protein GLY96|Immediate early response 3|Immediate early response 3 protein|Immediately early gene X 1|PACAP-responsive gene 1 protein|PRG 1|PRG1|Protein DIF 2|Protein DIF-2|Protein PRG1|Radiation-inducible immediate-early gene IEX-1
- DIF2| Differentiation-dependent gene 2 protein| GLY96| IER3| IEX1_HUMAN| IEX1L| Immediate early protein GLY96| Immediate early response 3| Immediate early response 3 protein| Immediately early gene X 1| PACAP-responsive gene 1 protein| PRG 1| PRG1| Protein DIF 2| Protein DIF-2| Protein PRG1| Radiation-inducible immediate-early gene IEX-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46695
- P46695
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Radiation-inducible immediate-early gene IEX-1 protein (1-82AA)
- Recombinant Human Radiation-inducible immediate-early gene IEX-1 protein (1-82AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together