Anti-KCNC4 Rabbit Polyclonal Antibody
Catalog # 77740-104
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KCNC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q03721
- Antigen Synonyms:C1orf30| Potassium voltage-gated channel subfamily C member 4| KSHIIIC| Voltage-gated potassium channel subunit Kv3.4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Voltage-gated potassium channel subunit Kv3.4
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: KCNC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse