Anti-ERLIN1 Rabbit Polyclonal Antibody
Catalog # 77736-127
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ERLIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O75477
- Antigen Synonyms:Band 7 23-211 Keo4 (Interim) similar to C.elegans protein| C10orf69| chromosome 10 open reading frame 69 | Endoplasmic reticulum lipid raft-associated protein 1| ER lipid raft associated 1 | Erlin 1| Erlin-1| ERLN1_HUMAN| KE04| KEO4| OTTHUMP00000059425| Protein KE04| SPFH domain containing protein 1| SPFH domain family| member 1| SPFH domain-containing protein 1| SPFH1| SPG62| Stomatin-prohibitin-flotillin-HflC/K domain containing protein 1| Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ERLIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat