Anti-NECAB3 Rabbit Polyclonal Antibody
Catalog # 77744-266
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NECAB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96P71
- Antigen Synonyms:amyloid beta (A4) precursor protein binding|family A|member 2 binding protein|Amyloid beta A4 protein binding family A member 2 binding protein|Amyloid beta A4 protein-binding family A member 2-binding protein|APBA2BP|dJ63M2.4|dJ63M2.5|EFCBP3|N terminal EF hand calcium binding protein 3|N-terminal EF-hand calcium-binding protein 3|NECA3_HUMAN|Nek2 interacting protein 1|Nek2-interacting protein 1|Neuronal calcium binding protein 3|Neuronal calcium-binding protein 3|NIP1|STIP3|Synaptotagmin interacting protein 2|SYTIP2|X11L binding protein 51|X11L-binding protein 51|XB51
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:XB51
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: NECAB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat