Anti-LY75 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LY75
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77859-246
- 77742-482
- Antigen Symbol:
- LY75
- LY75
- Antigen Synonyms:
- C-type lectin domain family 13 member B|CD 205|CD205|CD205 antigen|CLEC 13B|CLEC13B|DEC 205|DEC-205|DEC205|gp200|gp200 MR6|gp200-MR6|LY 75|Ly-75|LY75_HUMAN|Lymphocyte antigen 75|Renal Cell Carcinoma (gp200)
- C-type lectin domain family 13 member B|CD 205|CD205|CD205 antigen|CLEC 13B|CLEC13B|DEC 205|DEC-205|DEC205|gp200|gp200 MR6|gp200-MR6|LY 75|Ly-75|LY75_HUMAN|Lymphocyte antigen 75|Renal Cell Carcinoma (gp200)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60449
- O60449
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Lymphocyte antigen 75 protein (440-680AA)
- Recombinant human Lymphocyte antigen 75 protein (440-680AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together