Anti-IP6K2 Rabbit Recombinant polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: IP6K2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-162
- 77741-151
- Antigen Symbol:
- IP6K2
- IP6K2
- Antigen Synonyms:
- ATP 1D myo inositol hexakisphosphate phosphotransferase|IHPK 2|IHPK2|Inositol hexakisphosphate kinase 2|InsP6 kinase 2|IP6K2_HUMAN|KIAA0263|OTTHUMP00000164824|P(i) uptake stimulator|P(i)-uptake stimulator|Pi uptake stimulator|PiUS
- ATP 1D myo inositol hexakisphosphate phosphotransferase| IHPK 2| IHPK2| Inositol hexakisphosphate kinase 2| InsP6 kinase 2| IP6K2_HUMAN| KIAA0263| OTTHUMP00000164824| P(i) uptake stimulator| P(i)-uptake stimulator| Pi uptake stimulator| PiUS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHH9
- Q9UHH9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Inositol hexakisphosphate kinase 2 protein (1-180AA)
- Recombinant Human Inositol hexakisphosphate kinase 2 protein (1-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together