Anti-HIST1H1A Rabbit Polyclonal Antibody
New Product
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: HIST1H1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77739-702
- 77856-862
- Antigen Symbol:
- HIST1H1A
- HIST1H1A
- Antigen Synonyms:
- H1-1|H1F1|Histone H1.1|Histone H1a
- H1-1|H1F1|Histone H1.1|Histone H1a
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02539
- Q02539
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Peptide sequence around site of Lys (21) derived from Human Histone H1.1
- Peptide sequence around site of Lys (21) derived from Human Histone H1.1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together