Order Entry
United States
ContactUsLinkComponent
Anti-NR0B1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NR0B1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
Anti-NR0B1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77744-898
  • 77744-897
  • Antigen Symbol:
  • NR0B1
  • NR0B1
  • Antigen Synonyms:
  • AHC|AHCH|AHX|DAX 1|DAX1|Dosage sensitive sex reversal|DSS|DSS AHC critical region on the X chromosome protein 1|DSS-AHC critical region on the X chromosome protein 1|GTD|HHG|NR0B1_HUMAN|NROB1|Nuclear hormone receptor|Nuclear receptor 0B1|Nuclear receptor DAX 1|Nuclear receptor DAX-1|Nuclear receptor DAX1|Nuclear receptor subfamily 0 group B member 1|SRXY2
  • AHC|AHCH|AHX|DAX 1|DAX1|Dosage sensitive sex reversal|DSS|DSS AHC critical region on the X chromosome protein 1|DSS-AHC critical region on the X chromosome protein 1|GTD|HHG|NR0B1_HUMAN|NROB1|Nuclear hormone receptor|Nuclear receptor 0B1|Nuclear receptor DAX 1|Nuclear receptor DAX-1|Nuclear receptor DAX1|Nuclear receptor subfamily 0 group B member 1|SRXY2
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P51843
  • P51843
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Nuclear receptor subfamily 0 group B member 1 protein (1-243AA)
  • Nuclear receptor subfamily 0 group B member 1 protein (1-243AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together