Anti-FOXH1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: FOXH1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77855-552
- 77737-426
- Antigen Symbol:
- FOXH1
- FOXH1
- Antigen Synonyms:
- FAST 1|Fast 2|Fast-1|Fast-2|FAST1|Fast2|Forkhead activin signal transducer 1|Forkhead activin signal transducer 2|Forkhead box H1|Forkhead box protein H1|FOX H1|FOXH 1|FOXH1_HUMAN|hFAST 1|hFAST-1|hFAST1|Human homolog of Xenopus forkhead activin signal transducer 1|TGF beta/Activin signal transducer FAST1
- FAST 1| Fast 2| Fast-1| Fast-2| FAST1| Fast2| Forkhead activin signal transducer 1| Forkhead activin signal transducer 2| Forkhead box H1| Forkhead box protein H1| FOX H1| FOXH 1| FOXH1_HUMAN| hFAST 1| hFAST-1| hFAST1| Human homolog of Xenopus forkhead activin signal transducer 1| TGF beta/Activin signal transducer FAST1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75593
- O75593
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Forkhead box protein H1 protein (153-269AA)
- Recombinant Human Forkhead box protein H1 protein (153-269AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together