Anti-CABP7 Rabbit Polyclonal Antibody
Catalog # 77735-957
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CABP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q86V35
- Antigen Synonyms:CaBP7|CABP7_HUMAN|Calcium binding protein CaBP7|Calcium-binding protein 7|CALN2|Calneuron 2|Calneuron II|Calneuron-2|MGC129428|MGC129429|MGC57793|MGC94082|OTTMUSP00000005262|rCaBP7|RP23-280J3.1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:RP23-280J3.1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: CABP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat