Anti-HOXD9 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXD9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77740-338
- 77857-618
- Antigen Symbol:
- HOXD9
- HOXD9
- Antigen Synonyms:
- homeobox D9|Homeobox protein Hox D9|Homeobox protein Hox-4C|Homeobox protein Hox-5.2|Homeobox protein Hox-D9|Hox 4.3|Hox 5.2|HOX4|HOX4C|HXD9_HUMAN|RP23 313J15.10
- homeobox D9|Homeobox protein Hox D9|Homeobox protein Hox-4C|Homeobox protein Hox-5.2|Homeobox protein Hox-D9|Hox 4.3|Hox 5.2|HOX4|HOX4C|HXD9_HUMAN|RP23 313J15.10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P28356
- P28356
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-D9 protein (38-276AA)
- Recombinant Human Homeobox protein Hox-D9 protein (38-276AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together