Anti-IL17RB Rabbit Polyclonal Antibody
Catalog # 77740-956
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IL17RB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NRM6
- Antigen Synonyms:CRL4| EVI27| IL17BR| UNQ2501/PRO19612| Interleukin-17 receptor B| IL-17 receptor B| IL-17RB| Cytokine receptor-like 4| IL-17 receptor homolog 1| IL-17Rh1| IL17Rh1| Interleukin-17B receptor| IL-17B receptor
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:IL-17B receptor
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IL17RB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human