Anti-PITX3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PITX3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-733
- 77740-495
- Antigen Symbol:
- PITX3
- PITX3
- Antigen Synonyms:
- Homeobox protein PITX 3|Homeobox protein MGC12766|Paired like homeodomain transcription factor 3|Paired-like homeodomain transcription factor 3|Pituitary homeobox 3|PITX 3|PITX3_HUMAN|PTX 3|PTX3
- Homeobox protein PITX 3|Homeobox protein MGC12766|Paired like homeodomain transcription factor 3|Paired-like homeodomain transcription factor 3|Pituitary homeobox 3|PITX 3|PITX3_HUMAN|PTX 3|PTX3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75364
- O75364
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Pituitary homeobox 3 protein (10-200AA)
- Recombinant Human Pituitary homeobox 3 protein (10-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together