Anti-ETV4 Rabbit Polyclonal Antibody
77736-248
New Product
- Antigen Symbol:ETV4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P43268
- Antigen Synonyms:Adenovirus E1A enhancer binding protein|Adenovirus E1A enhancer-binding protein|E1A F|E1A-F|ETS translocation variant 4|ets variant gene 4 (E1A enhancer binding protein E1AF)|ETV4_HUMAN|PEA3|PEA3 protein|PEAS3|Polyomavirus enhancer activator 3|Polyomavirus enhancer activator 3 homolog|Protein PEA3
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Protein PEA3
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: ETV4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat