Anti-MYH7B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MYH7B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-741
- 77860-205
- Antigen Symbol:
- MYH7B
- MYH7B
- Antigen Synonyms:
- Antigen MLAA-21|cardiac muscle beta isoform|Myosin cardiac muscle beta chain|Myosin heavy chain 7B|Slow A MYH14|MYH7B_HUMAN|Myosin-7B
- Antigen MLAA-21|cardiac muscle beta isoform|MYH7B_HUMAN|Myosin cardiac muscle beta chain|Myosin heavy chain 7B|Myosin-7B|Slow A MYH14
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A7E2Y1
- A7E2Y1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Myosin-7B protein (1-176AA)
- Recombinant Human Myosin-7B protein (1-176AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together